missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIRC8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20144-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Description
Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family including IAP, XIAP/ILP-1/MIHA, and Livin/KIAP that bind to and inhibits specific proteases. A novel member in the IAP protein family was recently identified and designated ILP-2 for IAP-like protein-2. ILP-2 has high homology to ILP-1, but is encoded by a distinct gene that is solely expressed in testis of tested normal human tissues. ILP-2, unlike ILP-1, has no inhibitory effect on Fas and TNF induced apoptosis, but potently inhibits apoptosis induced by overexpression of Bax or by coexpression of caspase-9 with Apaf-1. ILP-2 interacts with the processed caspase-9. These results suggest that ILP-2 is a novel IAP family member with restricted specificity for caspase-9.Specifications
BIRC8 | |
Polyclonal | |
Unconjugated | |
BIRC8 | |
BIRC8, hILP2, IAP like protein 2, ILP 2, ILP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112401, 11798 | |
-20°C | |
Liquid |
Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q60989, Q96P09 | |
BIRC8, Xiap | |
BIRC8 Fusion Protein Ag13658 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |