Learn More
Uvex Black/Blue PU 85552 Boot - Width 11
Uvex 1 safety shoes protect and support the natural movement of the human body by reducing pressure and strain; protection classes S1 and S2
Marca:  Uvex 8555236
Dettagli aggiuntivi : Peso : 0.93400kg
Codice prodotto. 17235331
  • 148.00€ / 1 paio
Spedizione stimata: 09-11-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Uvex 1 Boot 8555.2 S3 SRC
  • Sporty, multifunctional safety boots
  • Especially lightweight and flexible
  • 100% metal-free Uvex Xenova™ toecap
  • Compact design, anatomical shape, good side stability, no thermal conductivity and anti-magnetic properties
  • Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
  • Made with new developed last technology for extended wearing comfort
  • Uvex Climazone technology ensures optimum wearer comfort
  • Breathable, water-resistant, leather upper material
  • Distance mesh lining provides optimum breathability
  • Virtually seamless design to eliminate pressure points
  • Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
  • Soft padded collar and tongue
  • Outstanding dual density polyurethane sole unit with best shock absorption characteristics and superb slip resistance
    • Free of lacquer wetting inhibitors such as silicones, phthalates, etc.
  • Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 8555.2
  • Standard EN ISO 20345:2011 S3 SRC
  • Sizes: 35 to 52
Specifica
Specifica
Blue, Black
36
Composite
EN ISO 20345:2011
Standard width
Yes
S3
Leather
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 1
No
SRC