missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLIMP1/PRDM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP2-27180SS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
BLIMP1/PRDM1 Polyclonal specifically detects BLIMP1/PRDM1 in Human, Bovine samples. It is validated for Western Blot.
Specifica
| BLIMP1/PRDM1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| It is 100% homologous in isoforms 1 and 2 of human BLIMP-1 (PRDM1). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| PRDM1 | |
| Amino acids 692-719 of human BLIMP-1 (PRDM1) were used as the immunogen for this antibody. | |
| 0.025 mL | |
| Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger | |
| 639 | |
| Human, Bovine | |
| Antisera |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto