Learn More
BLIMP1/PRDM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
240.00€ - 422.00€
Specifica
| Antigene | BLIMP1/PRDM1 |
|---|---|
| Diluizione | Western Blot 1:500-1:2000 |
| Applicazioni | Western Blot |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18149583
|
Novus Biologicals
NBP2-27180SS |
0.025 mL |
240.00€
25µL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18164635
|
Novus Biologicals
NBP2-27180 |
0.1 mL |
422.00€
0.10mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
BLIMP1/PRDM1 Polyclonal specifically detects BLIMP1/PRDM1 in Human, Bovine samples. It is validated for Western Blot.Specifica
| BLIMP1/PRDM1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Chromatin Research, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Zinc Finger | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| 639 | |
| Amino acids 692-719 of human BLIMP-1 (PRDM1) were used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Bovine | |
| Beta-interferon gene positive regulatory domain I-binding factor, beta-interferon gene positive-regulatory domain I binding factor, BLIMP-1, BLIMP1MGC118925, blmp-1, B-lymphocyte-induced maturation protein 1, MGC118922, MGC118923, Positive regulatory domain I-binding factor 1, PR domain containing 1, with ZNF domain, PR domain zinc finger protein 1, PR domain-containing protein 1, PRDI-BF1MGC118924, PRDI-binding factor 1, PRDI-binding factor-1, PR-domain zinc finger protein 1 | |
| PRDM1 | |
| IgG | |
| Unpurified | |
| It is 100% homologous in isoforms 1 and 2 of human BLIMP-1 (PRDM1). |
For Research Use Only