missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BMP61 M611 Self-Laminating Polyester Labels

A clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.

Marca:  Brady™ 150858

Dettagli aggiuntivi : Peso : 0.11000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17100411

  • 73.50€ / pezzi 500

Spedizione stimata: 20-01-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Good smudge resistance, solvent resistance and high/low-temperature performance
  • Service temperature of -320 to 230° F (-196 to 110° C)
  • Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
Specifica

Specifica

Acrylic
Polyester
Printer Labels
BMP™61 Labeling System, BradyPrinter™ M611
1.125 x 0.5 in. (W x H)
1.125 in.
White
Thermal Transfer
Laboratory ID - Bottle, Slide, Tube & Vial
500/Pk.
0.0019 in.
0.5 in.
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali