Learn More
WTW™ Boron B Test Kits
Marca: WTW™ 252041
Dettagli aggiuntivi : Peso : 0.40000kg
Specifica
Azomethine | |
PhotoLab™ S12, PhotoLab™ spectral photometers | |
25 | |
Photometric |
For waste water, drinking water, sea water | |
Certificates | |
25 Tests | |
Boron (B) cell test kit |