Learn More
WTW™ Boron B Reagent Test Kit
With certificate for optimum precision.
Marca: WTW™ 250427
Dettagli aggiuntivi : Peso : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Descrizione
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifica
Modified Winkler method | |
For PhotoLab™ S12, PhotoLab™ Spectral photometers | |
60 | |
Photometric |
For waste water, drinking water | |
Certificates | |
0.050 - 0.800mg/I B |