Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Marca: Lenz Laborglasinstrumente™ 04030070
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Cylindrical shape with straight discharge tube
- With NS socket and PE stopper
- With solid glass plug
- With Screwthreade retaining device
- With scale
Specifica
1000 mL | |
Borosilicate Glass | |
Polyethylene |