Learn More
Trajan Scientific and Medical™ BPX50 GC Capillary Columns
Designed for range of EPA and pharmaceutical methods. Trajan Scientific and Medical BPX50 GC Capillary Columns are MS premium, low bleed phase columns.
Marca: Trajan Scientific and Medical™ 054772
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Mid-polarity column
- Inert
- Low bleed
- High temperature
- Ideal for a range of EPA methods and pharmaceutical applications
Specifica
BPX50 GC Capillary Column | |
Steroids and a Variety of Pharmaceutical Applications Analysis, Triazines/Herbicides, Drug Screening, Steroids and a Variety of Pharmaceutical Applications | |
1 μm | |
50% Phenyl Polysilphenylene-siloxane |
0.53 mm | |
80°C to 330/350°C | |
30 m | |
BPX50 |