Learn More
Brady™ BradyJet J2000 Color Label Printer for EU
The BradyJet J2000 Color Label Printer is a practical and easy to use, full color inkjet label printer that can print reliable, high resolution signs, facility and laboratory sample labels.
Marca: Brady™ 150159
1705.25 EUR valido fino al 2024-12-31
Usa il codice promo "21918" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 3.17500kg
Descrizione
The Brady™ BradyJet J2000 Color Label Printer works great in a multitude of industries, from lab to safety and more.
- Compact design means it can fit in tight spaces
- On-demand, mid-volume inkjet solution creates photo quality color labels in a single pass
- More color options than you'd ever need at up to 16 million
- Custom identification without the high cost of outsourced printing
- Contact our Brady™ Technical Support Team to schedule a free phone appointment
Specifica
Inkjet | |
264 mm | |
180 mm | |
389 mm | |
Non-sterile | |
J2000 Printer, EU Power Cord with AC Adapter, USB Cable, Quick Start Guide, Ink Cartridge, Material Roll for Printhead Alignment, USB Stick with Drivers and Manuals, Brady™ Workstation Basic Suite | |
J2000 | |
4800 dpi | |
Windows™ 7, Windows™ 8, Windows™ 10 | |
7 lb. | |
Barcode Labeling, Electrical and Laboratory Labeling, General Identification, Hazard Identification | |
110/240 V |
10.4 in. | |
7 in. | |
15.3 in. | |
Not Autoclavable | |
Multi-color | |
Chinese (Tradditional), Chinese (Simplified), English, French, German, Italian, Japanese, Korean, Spanish | |
63.5 mm/s | |
USB | |
15°C to 32°C | |
3.2 kg | |
1 Year |