missing translation for 'onlineSavingsMsg'
Learn More

Bromomalonaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 15434027
Click to view available options
Quantità:
1 g
5 g
50 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
2065-75-0
C3H3BrO2
150.959
MFCD00459999
SURMYNZXHKLDFO-UHFFFAOYSA-N
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
74945
2-bromopropanedial
C(=O)C(C=O)Br
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
2065-75-0
C3H3BrO2
150.959
MFCD00459999
SURMYNZXHKLDFO-UHFFFAOYSA-N
2-bromomalonaldehyde, bromomalonaldehyde, bromomalondialdehyde, propanedial, bromo, 2-bromomalondialdehyde, 2-bromo-1,3-propanedial, malonaldehyde, bromo, bromo-malonaldehyde, zlchem 220, 2-bromo-malonaldehyde
74945
2-bromopropanedial
C(=O)C(C=O)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Bromomalonaldehyde
Punto di fusione ∽145°C (decomposition)
Formula lineare BrCH(CHO)2
Quantità 1 g
Sensibilità Air Sensitive
Peso formulazione 150.96
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'