Learn More
Hirschmann™ Amber Glass Stoppers
Light weight amber colored hollow glass stopper for the protection of UV-sensitive media.
Marca: Hirschmann™ 8902386
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- Acc. to DIN 12 252
Specifica
Stopper | |
Glass | |
NS 29/32 |
1/Ea. | |
For tight sealing of standard ground vessels |