missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution, pH 8 EU Pharmacopoeia, Fisher Chemical™

Catalog Number 12900734
Click to view available options
Quantità:
1000 mL
Imballaggio:
Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Materiale o nome chimico Buffer solution
CAS 1336-21-6
pH 8
Quantità 1000 mL
Formula molecolare H5NO
Numero MDL MFCD00066650
Imballaggio Bottle
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Grado Eu. Ph. Reference
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'