missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BVES Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12920-1-AP-150UL
This item is not returnable.
View return policy
Description
Blood vessel epicardial substance (BVES), also designated Popeye protein 1, is a transmembrane protein that plays a role in cell-cell interactions and adhesion, specifically at tight junctions. BVES is composed of an extracellular amino-terminus, three transmembrane domains and a cytoplasmic carboxyl-terminus. It is expressed in the developing coronary vascular system, specifically in the proepicardium, migrating epithelial epicardium, delaminated vasculogenic mesenchyme and vascular smooth muscle cells, where it functions to direct development in heart, epithelial and muscle cells during embryogenesis. BVES accumulates at points of cell-cell contact, such as filopodia and cell borders, and promotes adhesion prior to the arrival of E-cadherin.Specifications
| BVES | |
| Polyclonal | |
| Unconjugated | |
| BVES | |
| BVES, HBVES, POP1, POPDC1, Popeye protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11149, 23828, 365603 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3BCU4, Q8NE79, Q9ES83 | |
| Bves | |
| BVES Fusion Protein Ag3593 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction