missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Kit/CD117 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18696-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
KIT may be involved in testicular germ cell tumor (TGCT). A common maligcy in males representing 95% of all testicular neoplasms. TGCTs have various pathologic subtypes including: unclassified intratubular germ cell neoplasia, seminoma (including cases with syncytiotrophoblastic cells), spermatocytic seminoma, embryonal carcinoma, yolk sac tumor, choriocarcinoma, and teratoma. Note=The gene represented in this entry may be involved in disease pathogenesis.Specifica
c-Kit/CD117 | |
Polyclonal | |
Unconjugated | |
KIT | |
C Kit, CD117, KIT, PBT, Proto oncogene c Kit, SCFR, Tyrosine protein kinase Kit | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3815 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10721 | |
KIT | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |