Learn More
C14orf126 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551516
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: WVEVQRGLVI YVCFFKGADK ELLPKMVNTL LNVKLSETEN GKHVSILDLP GNILIIPQAT LGGRLKGRNM QYHSNSGKEE GFELYSQFVT LCEKEVAANS KCAEARVVVE HGTYGNRQVL KLDTNGPFTH LIFE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Deacylates mischarged D-aminoacyl-tRNAs. Also deacylates mischarged glycyl-tRNA(Ala), protecting cells against glycine mischarging by AlaRS. Probably acts by rejecting L-amino acids from its binding site rather than specific recognition of D-amino acids. Catalyzes the hydrolysis of D-tyrosyl-tRNA(Tyr), has no activity on correctly charged L-tyrosyl-tRNA(Tyr). By recycling D-aminoacyl-tRNA to D-amino acids and free tRNA molecules, this enzyme counteracts the toxicity associated with the formation of D-aminoacyl-tRNA entities in vivo and helps enforce protein L-homochirality. In contrast to DTD1, deacylates L-Ala mischarged on tRNA(Thr)(G4.U69) by alanine-tRNA ligase AARS (PubMed:29410408). Can deacylate L-Ala due to a relaxed specificity for substrate chirality caused by the trans conformation of the Gly-Pro motif in the active site (PubMed:29410408). Also hydrolyzes correctly charged, achiral, glycyl-tRNA(Gly) in vitro, although in vivo EEF1A1/EF-Tu may protect cognate achiral glycyl-tRNA(Gly) from DTD2-mediated deacetylation. [UniProt]Specifica
C14orf126 | |
Polyclonal | |
Unconjugated | |
DTD2 | |
4930578F06Rik; 6530401N04Rik; Animalia-specific tRNA deacylase; ATD; B830049N13Rik; C14orf126; D-aminoacyl-tRNA deacylase 2; DTD2; D-tyrosyl-tRNA deacylase 2; D-tyrosyl-tRNA deacylase 2 (putative); D-tyrosyl-tRNA(Tyr) deacylase 2; L-alanyl-tRNA deacylase; probable D-tyrosyl-tRNA(Tyr) deacylase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
112487 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96FN9 | |
DTD2 | |
Recombinant protein corresponding to Human C14orf126 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only