Learn More
C16orf70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20602-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The MW of this protein is 48 kDa, and our antibody specifically recognizes the 48 kDa protein.
The function of C20orf30 has not been widely studied, and is yet to be fully elucidated.Specifica
| C16orf70 | |
| Polyclonal | |
| Unconjugated | |
| C16orf70 | |
| C16orf6, C16orf70, lin 10, LIN10, UPF0183 protein C16orf70 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102547323, 80262 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BSU1 | |
| C16orf70, LOC102547323 | |
| C16orf70 Fusion Protein Ag14576 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |