missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11602-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This antibody is raised against C1qA which is the A-chain polypeptide of human complement subcomponent C1q.
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.Specifica
C1qA | |
Polyclonal | |
Unconjugated | |
C1QA | |
C1qA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12259, 298566, 712 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02745, P31720, P98086 | |
C1QA | |
C1qA Fusion Protein Ag2141 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |