Learn More
C21orf63 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553370
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: NKTVCEDQEL KLHCHESKFL NIYSATYGRR TQERDICSSK AERLPPFDCL SYSALQVLSR RCYGKQRCKI IVNNHHFGSP CLPGVKKYLT VTYACVPKNI LTAIDPAIAN LKPSLKQKDG EYGINFDPSG SKVLRKDGIL VSNSLA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
C21orf63 encodes a protein that binds heparin.Specifica
C21orf63 | |
Polyclonal | |
Unconjugated | |
EVA1C | |
1700092M14Rik; 4931408A02Rik; B18; B19; C21; C21orf63; C21orf64; DC42; EVA1; EVA-1; eva-1 homolog C; eva-1 homolog C (C. elegans); EVA1C; Fam176c; family with sequence similarity 176, member C; FLJ12894; IRA1; PRED34; Protein eva-1 homolog C; protein FAM176C; RGD1307569; SUE21; TBLR1; UNQ2504/PRO5993 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P58658 | |
EVA1C | |
Recombinant protein corresponding to Human C21orf63 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only