missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C5b-9 Antibody (aE11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Marca: Novus Biologicals NBP1-05120-0.025MG
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
C5b-9 Monoclonal specifically detects C5b-9 in Human, Porcine, Bacteria, Equine, Baboon samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Western Blot (Negative).
Specifica
| C5b-9 | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Western Blot (Negative) | |
| MAC, Membrane attack complex, sC5-b9, sC5b-9, TCC, Terminal complement complex | |
| Purified C5b-9 | |
| 0.025 mg | |
| Primary | |
| Equine, Baboon, Pig, Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), ELISA, Immunofluorescence, Immunocytochemistry | |
| aE11 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Binds both membrane-bound MAC and fluid phase SC5b-9 complexes. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto