Learn More
C5orf22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560225
Additional Details : Peso : 0.02000kg
Description
Immunogen sequence: IWFHPTWAQQ IREGRHHFLV GKDTSTTTIR VTSTDHYFLS DGLYVPEDQL ENQKPLQLDV IMVKPYKLCN NQEENDAVSS Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 86%.
C5orf22 Polyclonal antibody specifically detects C5orf22 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
C5orf22 | |
Polyclonal | |
Unconjugated | |
C5orf22 | |
C5orf22; chromosome 5 open reading frame 22; UPF0489 protein C5orf22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55322 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q49AR2 | |
C5orf22 | |
Recombinant protein corresponding to Human C5orf22 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only