Learn More
WTW™ Cadmium Reagent Test Kits
With certificate for optimum precision.
Brand: WTW™ 252051
Additional Details : Peso : 0.80000kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Description
- Convenient and cost-effective
- Assured quality by AQA/IQC
- For low concentrations and routine work with large sample throughput in the laboratory
- Wide measuring range using rectangular cuvettes for determination of trace concentrations
Specifications
Rhodamine B | |
For PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, spectroFlex photometers | |
55 | |
55 Tests | |
CombiCheck Gold (Au) reagent test kit |
For waste water, seepage water, process water | |
Certificates | |
0.002 - 0.500mg/L | |
Photometric |