missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10492-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Calpain, a heterodimer consisting of a large and a small subunit, is a major intracellular protease, although its function has not been well established. This gene encodes a muscle-specific member of the calpain large subunit family that specifically binds to titin. Mutations in this gene are associated with limb-girdle muscular dystrophies type 2A. Alternate promoters and alternative splicing result in multiple transcript variants encoding different isoforms and some variants are ubiquitously expressed.Specifica
Calpain 3 | |
Polyclonal | |
Unconjugated | |
CAPN3 | |
Calpain 3, calpain 3, (p94), Calpain L3, Calpain p94, CANP 3, CANP3, CANPL3, CAPN3, LGMD2, LGMD2A, nCL 1, NCL1, New calpain 1, p94 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12335, 29155, 825 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P16259, P20807, Q64691 | |
Capn3 | |
Calpain 3 Fusion Protein Ag0760 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |