Learn More
Motic™ Camera Adapter, C-Mount, B3
Choose the most appropriate adapter for your camera and microscope and unlock the potential of photomicography. Motic™ Camera Adapter, C-Mount, B3 , is suitable for B3 Series Microscopes and uses the industry standard attachment for digital imaging devices dedicated to microscopes.
Marca: Motic™ SW0113M9
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Connect a C-mount camera with this adapter.
- Suitable for B3 Series Microscopes.
- B3 Microscopes are designed for advanced laboratory work and employ ASC achromatic super contrast and plan achromatic objectives.
Specifica
C-Mount Camera Adapter | |
B3 Microscopes, 1/2in. Sensor Camera | |
0.5X |