Learn More
Thermo Scientific™ Carrying Kit Set for Eutech™ CyberScan CON 11/110, CON 400/410
Eutech™ Carrying Kit Cases are thoughtfully designed to hold your Eutech™ meter and electrodes.
Marca: Thermo Scientific™ 01X266802
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
Eutech™ Carrying Kit Cases are thoughtfully designed to hold Eutech™ meters and electrodes.The Economy Carrying Kit Set is a plastic carrying case for CyberScan CON 11/110, CON 400/410 meters and probes, calibration solutions, batteries, and manuals.
The Carrying Kit set is comprised of:
- 1413 μS/CM KCL Calibration Solution
Specifica
CyberScan Conductivity/TDS Carrying Kit Set – Plastic Carrying Case comprises: 1 x 1413 mS, 12.88 mS KCl, 3000 ppm 442 standard solutions and 1 x deionised rinse water (60 ml) | |
Accessory | |
1413 μS/CM KCL Calibration Solution, 2.88 mS/cm KCl Calibration Solution, 3000 ppm 442 TDS standard solutions, deionised rinse water (60 ml) |