Learn More
3M™ Cartridge Filter Holder 7586
3M™ Cartridge Filter Holder is for 3M™ 7500 Series Half Masks and includes two inhalation valves and one exhalation valve.
Marca: 3M™ 7000029733
20.85 EUR valido fino al 2024-12-31
Usa il codice promo "22912" per avere il tuo prezzo promozionale.
Additional Details : Peso : 0.13500kg
Beschreibung
- Replacement cartridge filter holder for half mask respirator
- Pack includes two inhalation valves and one exhalation valve
- For use with 3M™ Half Face Masks 7500 Series
Spezifikation
CE Approved | |
Gray | |
Cartridge Filter Holder | |
Two inhalation valves and one exhalation valve |