Learn More
Thermo Scientific™ Caspase-8 Inhibitor I, cell-permeable
Marca: Thermo Scientific™ J66050.M¬
Dettagli aggiuntivi : Peso : 0.00001kg
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
1 mg | |
-30°C to -10°C | |
2000.41g/mol | |
Signal Transduction Reagent |
White to Off-white | |
C95H162N20O26 | |
N-Acetyl-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Ile-Glu-Thr-Asp-al; Ac-AAVALLPAVLLALLAPIETD-CHO | |
Lyophilized Powder |