Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Marca:  Fisherbrand™
Dettagli aggiuntivi : Peso : 0.04000kg
Codice prodotto. 15561375
  • 124.00€ / pezzi 6
Spedizione stimata: 20-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Preconfigured ready-to-use links and assemblies.
  • Time-saving assembly
  • Four formulations for range of chemical compatibility
  • Use with Catalyst extension coils/tubing
Specifica
Specifica
Stopped tubing assembly
2.79 mm