missing translation for 'onlineSavingsMsg'
Learn More

CBLB Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D3A2, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66353-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898145

  • 325.38€ / 150µL

Spedizione stimata: 06-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.
Specifica

Specifica

CBLB
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13191, Q3TTA7, Q8K4S7
Cblb
CBLB Fusion Protein Ag18757
150 μL
Primary
Rat, Porcine, Mouse, Human
Antibody
IgG3
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
2D3A2
Unconjugated
CBLB
CBLB, Nbla00127, RING finger protein 56, RNF56, SH3 binding protein CBL B
Mouse
Protein A
RUO
100621690, 171136, 208650, 868
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali