Learn More
CCDC90B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA552876
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MNSRQAWRLL LSQGRGDRWV SRPRGHFSPA LRREFFTTTT KEGYDRRPVD ITPLEQRKLT FDTHALVQDL ETHGFDKTQA ETIVSALTAL SNVSLDTIYK EMVTQAQQEI TVQQL Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 75%.
CCDC90B Polyclonal antibody specifically detects CCDC90B in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifica
CCDC90B | |
Polyclonal | |
Unconjugated | |
CCDC90B | |
2310015N07Rik; C86558; CCDC90B; coiled-coil domain containing 90B; coiled-coil domain-containing protein 90B, mitochondrial; CUA003; MDS011; MDS025; RGD1308637 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
60492 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZT6 | |
CCDC90B | |
Recombinant protein corresponding to Human CCDC90B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only