Learn More
CCR7 (CD197) Mouse anti-Human, Brilliant Violet 421, Clone: 2-L1-A, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566743
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.
Specifica
| CCR7 (CD197) | |
| Monoclonal | |
| Brilliant Violet 421 | |
| 100 Tests | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 2-L1-A | |
| Mouse | |
| RUO | |
| CCR7 (CD197) | |
| Refrigerate (2 to 8°C) |