Learn More
CD11c Mouse, anti-Human, R718, Clone: B-ly6, BD Horizon™
Mouse Monoclonal Antibody
Marca:  BD Biosciences 566933
Dettagli aggiuntivi : Peso : 0.00020kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17227724
  • 199.00€ / 25 tests
Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
Specifica
Specifica
CD11c
Monoclonal
R718
25 Tests
Primary
Refrigerate Upon Arrival
Flow Cytometry
B-ly6
Mouse