missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Mouse anti-Human, BUV563, Clone: GHI/61, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 741402
Dettagli aggiuntivi : Peso : 0.00030kg
Descrizione
CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for Flow CytometrySpecifica
CD163 | |
Monoclonal | |
Brilliant Ultraviolet 563 | |
CD163 antigen; M130; MM130; GHI/61; D11C163A; RM3/1 | |
Glycoprotein preparation from Hairy Cell Leukemia Spleen | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
GHI/61 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody. |