Learn More
CD19 Mouse anti-Human, BB700, Clone: SJ25C1 (also known as SJ25-C1), BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566397
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Specifica
| CD19 | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
| Flow Cytometry | |
| SJ25C1 (also known as SJ25-C1) | |
| Mouse | |
| RUO | |
| Human | |
| Purified |