missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1a Mouse anti-Human, BUV737, Clone: HI149, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 748853
Dettagli aggiuntivi : Peso : 0.00030kg
Descrizione
CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Flow CytometrySpecifica
CD1a | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
CD1; T6/Leu-6; R4; T6; HTA-1; FCB6 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Flow Cytometry | |
HI149 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG1 κ |