missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human, BUV737, Clone: L128, BD Horizon™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612830
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow Cytometry.Specifica
CD27 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
25 Tests | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
L128 | |
Mouse | |
RUO | |
CD27 | |
Refrigerate (2 to 8°C) |