missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human , Alexa Fluor 647, Clone: OKT4, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 566681
Dettagli aggiuntivi : Peso : 0.00060kg
Descrizione
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifica
CD4 | |
Monoclonal | |
Alexa Fluor 647 | |
CD4 antigen (p55); T-cell surface antigen T4/Leu-3 | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
RUO | |
Human | |
Mouse BALB/c x A/J, also known as CAF1 IgG2b κ |
Flow Cytometry | |
OKT4 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor 647 under optimum conditions, and unreacted Alexa Fluor 647 was removed. |