missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Mouse anti-Human , BUV615, Clone: SK3 (also known as Leu3a), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 612988
Dettagli aggiuntivi : Peso : 0.00020kg
Descrizione
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow CytometrySpecifica
CD4 | |
Monoclonal | |
Brilliant Ultraviolet 615 | |
L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55) | |
Human Peripheral Blood T Cells | |
RUO | |
Human | |
Mouse BALB/c IgG1 κ |
Flow Cytometry | |
SK3 (also known as Leu3a) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |