missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40 / TNFRSF5 / CD40L-Receptor Mouse, Clone: C40/2383, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 958-MSM3-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD40 / TNFRSF5 / CD40L-Receptor | |
Cell Surface and Secreted | |
C40/2383 | |
Unconjugated | |
Mouse | |
43kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
958 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human CD40 protein (apprx. aa 29-107) | |
L-428 Cells. Tonsil, Spleen or Thymus | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |