missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (H5C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-42225SS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD63 Monoclonal antibody specifically detects CD63 in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay, In vitro Assay, Dot Blot, Electron Microscopy, Flow (Intracellular), Immunohistochemistry (Whole-Mount), Neutralization
Specifica
| CD63 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry 1:1000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunoprecipitation, Functional reported in scientific literature (PMID 9811687), In vitro assay reported in scientific literature (PMID 21464080), Dot Blot, Electron Microscopy reported in scientific literature (PMID 16735575), Flow (Intracellular), Immunohistochemistry Whole-Mount reported in scientific literature (PMID 21464080), Block/Neutralize reported in scientific literature (PMID 35602933) | |
| P08962 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| H5C6 | |
| Unconjugated | |
| PBS | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| Human splenic adherent cells. | |
| 0.025 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 967 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto