missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (H5C6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
237.00€ - 657.00€
Specifica
| Antigene | CD63 |
|---|---|
| Clone | H5C6 |
| Concentrazione | 1.0 mg/mL |
| Diluizione | Western Blot, Flow Cytometry 1:1000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunoprecipitation, Functional reported in scientific literature (PMID 9811687), In vitro assay reported in scientific literature (PMID 21464080), Dot Blot, Electron Microscopy reported in scientific literature (PMID 16735575), Flow (Intracellular), Immunohistochemistry Whole-Mount reported in scientific literature (PMID 21464080), Block/Neutralize reported in scientific literature (PMID 35602933) |
| Applicazioni | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
18682668
|
Novus Biologicals
NBP2-42225 |
0.1 mL |
657.00€
0.10mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
18600318
|
Novus Biologicals
NBP2-42225SS |
0.025 mL |
237.00€
0.03mL |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
CD63 Monoclonal antibody specifically detects CD63 in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay, In vitro Assay, Dot Blot, Electron Microscopy, Flow (Intracellular), Immunohistochemistry (Whole-Mount), NeutralizationSpecifica
| CD63 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| PBS | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| Human splenic adherent cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| H5C6 | |
| Western Blot, Flow Cytometry 1:1000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunoprecipitation, Functional reported in scientific literature (PMID 9811687), In vitro assay reported in scientific literature (PMID 21464080), Dot Blot, Electron Microscopy reported in scientific literature (PMID 16735575), Flow (Intracellular), Immunohistochemistry Whole-Mount reported in scientific literature (PMID 21464080), Block/Neutralize reported in scientific literature (PMID 35602933) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Canine | |
| P08962 | |
| 967 | |
| IgG1 κ | |
| Protein G purified |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto