Learn More
CD8 Mouse anti-Human, Unconjugated, Clone: 1G2B10, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66868-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The CD8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD8. This antibody recognizes human antigen. The CD8 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifica
| CD8 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1G2B10 | |
| Unconjugated | |
| 150 μL | |
| 925 | |
| IgG1 |