missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8a Rat, V450, Clone: 53-6.7, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB16-V450-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CD8a | |
Monoclonal | |
0.2mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Cd8a | |
Protein A/G Purified | |
RUO | |
12525 | |
IgG2a κ |
Flow Cytometry | |
53-6.7 | |
V450 | |
NP_033987 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |