missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19794-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody is raised against the C-term of CDH10.
CDH10 is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein′s homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is predomitly expressed in brain and is putatively involved in synaptic adhesions, axon outgrowth and guidance.Specifica
CDH10 | |
Polyclonal | |
Unconjugated | |
CDH10 | |
Cadherin 10, CDH10, T2 cadherin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1008, 29181, 320873 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70408, Q9Y6N8 | |
Cdh10 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |