missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567791
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Predicted to react with Mouse samples.
CDH12 is a type II classical cadherin from the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin appears to be expressed specifically in the brain and its temporal pattern of expression would be consistent with a role during a critical period of neuronal development, perhaps specifically during synaptogenesis.Specifica
CDH12 | |
Polyclonal | |
Unconjugated | |
CDH12 | |
Brain cadherin; brain-cadherin; Br-cadherin; Cadherin; cadherin 12; cadherin 12, type 2 (N-cadherin 2); cadherin, neural type, 2; cadherin-12; CDH12; CDHB; N-cadherin 2; neural type cadherin 2; neuronal cadherin 2; RGD1566350 | |
Rabbit | |
affinity chromatography | |
RUO | |
1010, 215654 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
P55289, Q5RJH3 | |
CDH12 | |
Synthesized peptide derived from human CDH12. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |