Learn More
Cefoperazone sodium salt, Thermo Scientific™
Cefoperazone sodium salt, CAS # 62893-20-3, is the sodium salt form of cefoperazone. It is a semi-synthetic, broad-spectrum, and third-generation cephalosporin compound with antibiotic and bactericidal activities.
Marca: Thermo Scientific Alfa Aesar J65185.03
| Quantità | 1g |
|---|
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Cefoperazone sodium salt is a semi-synthetic, broad-spectrum, third-generation cephalosporin compound with antibiotic and bactericidal activities
• This compound can inhibit bacterial cell wall synthesis by inactivating penicillin-binding proteins and consequently interfering with the final transpeptidation step required for cross-linking peptidoglycan units, an important component of bacterial cell walls. This leads to a reduction in cell wall stability, causing cell lysis
Application
• Cefoperazone sodium salt exhibits activity against β-lactamase resistant microorganisms
• This compound is also used for studying the expression, binding, and inhibition of penicillin-binding proteins
• Cefoperazone is active in vitro against most gram-positive and gram-negative bacteria, including β-lactamase strains
• This compound has the ability to inhibit ∝-antitrypsin
Identificatori chimici
| 62893-20-3 | |
| 670.672 | |
| IHWPDDMUNKOOFD-BYLXUVCXSA-N | |
| 131673981 | |
| [HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na] |
| C25H29N9NaO8S2 | |
| MFCD07793331 | |
| Cefobid sodium salt; Cefazone sodium salt | |
| (6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
Specifica
| Cefoperazone sodium salt | |
| White | |
| C25H29N9NaO8S2 | |
| 4902135 | |
| Cefobid sodium salt; Cefazone sodium salt | |
| IHWPDDMUNKOOFD-BYLXUVCXSA-N | |
| (6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium | |
| 131673981 |
| 62893-20-3 | |
| 1g | |
| MFCD07793331 | |
| 14,1930 | |
| Soluble in water; Slightly soluble in alcohol. | |
| [HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na] | |
| 670.672 | |
| 667.65 |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.