Learn More
Cefoperazone sodium salt, Thermo Scientific™
Cefoperazone sodium salt, CAS # 62893-20-3, is the sodium salt form of cefoperazone. It is a semi-synthetic, broad-spectrum, and third-generation cephalosporin compound with antibiotic and bactericidal activities.
87.59€ - 568.02€
Identificatori chimici
| CAS | 62893-20-3 |
|---|---|
| Formula molecolare | C25H29N9NaO8S2 |
| Molecular Weight (g/mol) | 670.672 |
| Numero MDL | MFCD07793331 |
| InChI Key | IHWPDDMUNKOOFD-BYLXUVCXSA-N |
| Sinonimo | Cefobid sodium salt; Cefazone sodium salt |
| PubChem CID | 131673981 |
| IUPAC Name | (6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
| SMILES | [HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na] |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
15406779
|
Thermo Scientific Alfa Aesar
J65185.03 |
1g |
89.40€ 87.59€ / 1g Risparmia 1.81€ 2% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15416779
|
Thermo Scientific Alfa Aesar
J65185.06 |
5g |
210.00€ 206.00€ / 5g Risparmia 4.00€ 1% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
|
15426779
|
Thermo Scientific Alfa Aesar
J65185.14 |
25g |
579.00€ 568.02€ / 25g Risparmia 10.98€ 1% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Cefoperazone sodium salt is a semi-synthetic, broad-spectrum, third-generation cephalosporin compound with antibiotic and bactericidal activities
• This compound can inhibit bacterial cell wall synthesis by inactivating penicillin-binding proteins and consequently interfering with the final transpeptidation step required for cross-linking peptidoglycan units, an important component of bacterial cell walls. This leads to a reduction in cell wall stability, causing cell lysis
Application
• Cefoperazone sodium salt exhibits activity against β-lactamase resistant microorganisms
• This compound is also used for studying the expression, binding, and inhibition of penicillin-binding proteins
• Cefoperazone is active in vitro against most gram-positive and gram-negative bacteria, including β-lactamase strains
• This compound has the ability to inhibit ∝-antitrypsin
Identificatori chimici
| 62893-20-3 | |
| 670.672 | |
| IHWPDDMUNKOOFD-BYLXUVCXSA-N | |
| 131673981 | |
| [HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na] |
| C25H29N9NaO8S2 | |
| MFCD07793331 | |
| Cefobid sodium salt; Cefazone sodium salt | |
| (6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium |
Specifica
| Cefoperazone sodium salt | |
| White | |
| MFCD07793331 | |
| 14,1930 | |
| Soluble in water; Slightly soluble in alcohol. | |
| [HH].CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=C(C=C2)O)C(=O)NC3C4N(C3=O)C(=C(CS4)CSC5=NN=NN5C)C(=O)O.[Na] | |
| 670.672 | |
| 667.65 |
| 62893-20-3 | |
| C25H29N9NaO8S2 | |
| 4902135 | |
| Cefobid sodium salt; Cefazone sodium salt | |
| IHWPDDMUNKOOFD-BYLXUVCXSA-N | |
| (6R,7R)-7-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-(4-hydroxyphenyl)acetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;molecular hydrogen;sodium | |
| 131673981 |
Sicurezza e movimentazione
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
H317-H334
missing translation for 'einecsNumber' : 263-751-5
missing translation for 'rtecsNumber' : XI0374000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.