missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CELA3B / ELA3B Mouse, Clone: CELA3B/1811, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 23436-MSM11-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CELA3B / ELA3B | |
Cytoplasmic | |
CELA3B/1811 | |
Unconjugated | |
NP_031378 | |
Mouse | |
17kDa | |
Protein A/G Purified | |
RUO | |
23436 | |
IgG2b κ |
Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CELA3B | |
Recombinant human CELA3B protein fragment (aa82-238) | |
Panc-28 or PANC1 cells. Pancreas | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |