Learn More
Macherey-Nagel™ Chromafix™ HR-XA Cartridge
Marca: Macherey-Nagel™ 731769
Dettagli aggiuntivi : Peso : 0.10000kg
Specifica
Spherical PS/DVB Copolymer | |
Acidic Active Ingredients From Strongly Matrix-Contaminated Samples Like, e.g., Urine, Plasma, Serum, Phenolic Acids, Acidic Herbicides | |
Strong quat. ammonium modified polymeric anion exchanger | |
55 to 65 Å | |
850 m2/g |
240 mg | |
45 or 85 μm | |
Chromafix HR-XA Cartridge | |
50 Pack |