Learn More
Chromalite™ A860/5952 Macroporous Polyacrylic Resin Crosslinked with Divinylbenzene Strong Anion Exchanger, Purolite™
Chromalite™ A860/5952 is a porous polyacrylic resin crosslinked with DVB. Chromalite™ A860/5952 is an ideal choice, due to its hydrophilic and protein-compatible matrix for food and especially diary applications.
Marca: Chromalite LS73082-94
Dettagli aggiuntivi : Peso : 0.71000kg
Descrizione
Ideal for isolation of proteins, especially for proteins from milk and dairy products.
- Optimal for lactoferrin purification from whey
- Hydrophilic, protein-compatible matrix
- High rigidity
- Suitable for use in industrial columns
- Compliant with ResAP(2004) 3
- Halal, Kosher and TSE/BSE/GMO free
Specifica
Spherical Beads | |
0.8 eq/L | |
A860/5952 Macroporous Polyacrylic Resin | |
1 to 14 |
300 to 850 μm | |
1 L | |
Quaternary Ammonium |